Bovine;Dog;Horse;Human;Mouse;Rabbit;Rat Osteostatin (1-5) amide Trifluoroacetate
Catalog #: H-2244.0025BA
Supplier: Bachem Americas
CAS Number:
Specifications
- Species:RabbitHorseHumanBovineDogRatMouse
- CAS No.:155918-12-0
- Size:25 mg
- Storage conditions:–20 ± 5 °C
- MDL No.:MFCD18782139
- Protein synonyms:dog|pTH-Related Protein (107-111) amide (human|bovine|rat)|mouse|Hypercalcemia of Malignancy Factor (107-111) amide (human|rabbit|pTH-rP (107-111) amide (human|horse
- Protein/peptide name:Osteostatin (1-5) amide Trifluoroacetate
- Molecular weight:618.69
- Sequence:H-Thr-Arg-Ser-Ala-Trp-NH₂
- Molecular formula:C₂₇H₄₂N₁₀O₇
- Cat. no.:H-2244.0025BA
- Supplier No.:H-2244.0025
Specifications
About this item
This pTHrP fragment stimulated membrane-associated protein kinase C in freshly isolated rat spleen lymphocytes and thus raises the possibility of being a physiological regulator of the proliferation and other activities of lymphocytes.
Customers Who Bought This Also Bought