Hydrin 1
Catalog #: H-1558.0001BA
Supplier: Bachem Americas
CAS Number:
Specifications
- CAS No.:148440-73-7
- Size:1 mg
- Storage conditions:–20 ± 5 °C
- MDL No.:MFCD00237115
- Protein synonyms:Lys11)-Vasotocin|Gly10|(Arg8
- Protein/peptide name:Hydrin 1
- Molecular weight:1236.44
- Sequence:H-Cys-Tyr-Ile-Gln-Asn-Cys-Pro-Arg-Gly-Gly-Lys-OH
- Molecular formula:C₅₁H₈₁N₁₇O₁₅S₂
- Cat. no.:H-1558.0001BA
- Supplier No.:H-1558.0001
Specifications
About this item
The amphibian skin is a rich source of antimicrobial peptides, please see the same-named product family Other amphibian peptides as bombesin, caerulein, deltorphin, dermorphin, pseudin, sauvagine, and xenin show considerable pharmacological activity.
The Xenopus peptide hydrin 1' shows equivalent aldosterone-releasing activity compared to hydrin 1 and is also effective in stimulating water flux from the isolated toad bladder.
Customers Who Bought This Also Bought