H-Gly-Asp-Gly-OH
Catalog #: H-1308.0250BA
Supplier: Bachem Americas
CAS Number:
Specifications
- CAS No.:10517-27-8
- Size:250 mg
- Storage conditions:–20 ± 5 °C
- MDL No.:MFCD00237903
- Protein/peptide name:H-Gly-Asp-Gly-OH
- Molecular weight:247.21
- Sequence:H-Gly-Asp-Gly-OH
- Molecular formula:C₈H₁₃N₃O₆
- Cat. no.:H-1308.0250BA
- Supplier No.:H-1308.0250
Specifications
About this item
GDG.
Customers Who Bought This Also Bought