Ac-Asp(Glu-OH)-OH
Supplier: Bachem Americas
Certificates
Certificates
About this item
A non-hydrolyzable NAAG isomer which acts as selective metabotropic glutamate receptor-3 (mGluR3) antagonist and NAAG peptidase inhibitor.
Customers Who Bought This Also Bought
Water, HiPerSolv CHROMANORM® for HPLC, VWR Chemicals BDH®
Supplier: VWR International
73,87 $ to 302,61 $
Nalgene® VERSI-DRY® Lab Table Soakers, Thermo Scientific
Supplier: Thermo Fisher Scientific
1 098,06 $
SP Bel-Art Covamat™ Clear Bench Liner, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
362,33 $
Pierce™ DNA Extraction Buffer, Phenol:Chloroform and Tris Buffer, Thermo Scientific
Supplier: Thermo Scientific
628,97 $
SP Bel-Art Fritware® Porous Polyethylene Sheets, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
464,49 $
Nalgene® Polyolefin Plastic Paper Sheets, Thermo Scientific
Supplier: Thermo Fisher Scientific
534,61 $ to 1 523,31 $