Ac-Phe-Lys-OH
Catalog #: G-4090.0001BA
Supplier: Bachem Americas
CAS Number:
Specifications
- CAS No.:14287-21-9
- Size:1 g
- Storage conditions:–20 ± 5 °C
- MDL No.:MFCD00083211
- Protein/peptide name:Ac-Phe-Lys-OH
- Molecular weight:335.40
- Sequence:Ac-Phe-Lys-OH
- Molecular formula:C₁₇H₂₅N₃O₄
- Cat. no.:G-4090.0001BA
- Supplier No.:G-4090.0001
Specifications
About this item
Inverted aspartame-type sweetener. Adrover et al. used Ac-Phe-Lys as a model compound in their studies on protein glycation.
Customers Who Bought This Also Bought