H-Leu-Asp-OH
Catalog #: G-3835.0001BA
Supplier: Bachem Americas
CAS Number:
Specifications
- CAS No.:32949-40-9
- Size:1 g
- Storage conditions:–20 ± 5 °C
- MDL No.:MFCD00190941
- Protein/peptide name:H-Leu-Asp-OH
- Molecular weight:246.26
- Sequence:H-Leu-Asp-OH
- Molecular formula:C₁₀H₁₈N₂O₅
- Cat. no.:G-3835.0001BA
- Supplier No.:G-3835.0001
Specifications
About this item
Customers Who Bought This Also Bought