H-Asp-Trp-OH, Bachem
Catalog #: G-3705.0250BA
Supplier: Bachem Americas
CAS Number:
Specifications
- CAS No.:71835-79-5
- Size:250 mg
- Storage conditions:–20 ± 5 °C
- MDL No.:MFCD00237825
- Protein/peptide name:H-Asp-Trp-OH
- Molecular weight:319.32
- Sequence:H-Asp-Trp-OH
- Molecular formula:C₁₅H₁₇N₃O₅
- Cat. no.:G-3705.0250BA
- Supplier No.:G-3705.0250
Specifications
About this item
Customers Who Bought This Also Bought