H-Val-Asp-OH
Catalog #: G-3510.0001BA
Supplier: Bachem Americas
CAS Number:
Specifications
- CAS No.:20556-16-5
- Size:1 g
- Storage conditions:–20 ± 5 °C
- MDL No.:MFCD00037280
- Protein/peptide name:H-Val-Asp-OH
- Molecular weight:232.24
- Sequence:H-Val-Asp-OH
- Molecular formula:C₉H₁₆N₂O₅
- Cat. no.:G-3510.0001BA
- Supplier No.:G-3510.0001
Specifications
About this item
Customers Who Bought This Also Bought