H-Thr-Phe-OH
Catalog #: G-3295.0001BA
Supplier: Bachem Americas
CAS Number:
Specifications
- CAS No.:16875-27-7
- Size:1 g
- Storage conditions:–20 ± 5 °C
- MDL No.:MFCD00038591
- Protein/peptide name:H-Thr-Phe-OH
- Molecular weight:266.30
- Sequence:H-Thr-Phe-OH
- Molecular formula:C₁₃H₁₈N₂O₄
- Cat. no.:G-3295.0001BA
- Supplier No.:G-3295.0001
Specifications
About this item
Customers Who Bought This Also Bought