H-Ser-Leu-OH
Catalog #: G-3200.0001BA
Supplier: Bachem Americas
CAS Number:
Specifications
- CAS No.:6665-16-3
- Size:1 g
- Storage conditions:–20 ± 5 °C
- MDL No.:MFCD00055934
- Protein/peptide name:H-Ser-Leu-OH
- Molecular weight:218.25
- Sequence:H-Ser-Leu-OH
- Molecular formula:C₉H₁₈N₂O₄
- Cat. no.:G-3200.0001BA
- Supplier No.:G-3200.0001
Specifications
About this item
Substrate for human kidney dipeptidase.
Customers Who Bought This Also Bought