Pz-Pro-Leu-OH
Catalog #: G-2970.0001BA
Supplier: Bachem Americas
CAS Number:
Specifications
- CAS No.:98640-71-2
- Size:1 g
- Storage conditions:–20 ± 5 °C
- MDL No.:MFCD00077156
- Protein/peptide name:Pz-Pro-Leu-OH
- Molecular weight:466.54
- Sequence:Pz-Pro-Leu-OH
- Molecular formula:C₂₅H₃₀N₄O₅
- Cat. no.:G-2970.0001BA
- Supplier No.:G-2970.0001
Specifications
About this item
See also M-2460 and M-2465.
Cleavage product of Pz-peptide, M-1715, a substrate for collagenase and thimet oligopeptidase.
Customers Who Bought This Also Bought