H-Phe-Asp-OH
Catalog #: G-2870.0001BA
Supplier: Bachem Americas
CAS Number:
Specifications
- CAS No.:22828-05-3
- Size:1 g
- Storage conditions:–20 ± 5 °C
- MDL No.:MFCD00037281
- Protein/peptide name:H-Phe-Asp-OH
- Molecular weight:280.28
- Sequence:H-Phe-Asp-OH
- Molecular formula:C₁₃H₁₆N₂O₅
- Cat. no.:G-2870.0001BA
- Supplier No.:G-2870.0001
Specifications
About this item
Customers Who Bought This Also Bought