H-Met-Asp-OH
Catalog #: G-2730.0001BA
Supplier: Bachem Americas
CAS Number:
Specifications
- CAS No.:14595-65-4
- Size:1 g
- Storage conditions:–20 ± 5 °C
- MDL No.:MFCD00037282
- Protein/peptide name:H-Met-Asp-OH
- Molecular weight:264.30
- Sequence:H-Met-Asp-OH
- Molecular formula:C₉H₁₆N₂O₅S
- Cat. no.:G-2730.0001BA
- Supplier No.:G-2730.0001
Specifications
About this item
Customers Who Bought This Also Bought