Ac-Lys-D-Ala-D-lactic Acid · Acetate
Catalog #: G-4410.0050BA
Supplier: Bachem Americas
CAS Number:
Specifications
- CAS No.:282729-62-8
- Size:50 mg
- Storage conditions:–20 ± 5 °C
- MDL No.:MFCD00672392
- Protein/peptide name:Ac-Lys-D-Ala-D-lactic acid · acetate
- Molecular weight:391.42
- Sequence:Ac-Lys-D-Ala-D-lactic acid · acetate
- Molecular formula:C₁₄H₂₅N₃O₆ · C₂H₄O₂
- Cat. no.:G-4410.0050BA
- Supplier No.:G-4410.0050
Specifications
About this item
See also F-3260.
Ac-Lys-D-Ala-D-Lac-OH, depsipeptide analog of H-8005.
Customers Who Bought This Also Bought