H-Lys-Asp-OH
Catalog #: G-2640.0001BA
Supplier: Bachem Americas
CAS Number:
Specifications
- CAS No.:20556-18-7
- Size:1 g
- Storage conditions:–20 ± 5 °C
- MDL No.:MFCD00038214
- Protein/peptide name:H-Lys-Asp-OH
- Molecular weight:261.28
- Sequence:H-Lys-Asp-OH
- Molecular formula:C₁₀H₁₉N₃O₅
- Cat. no.:G-2640.0001BA
- Supplier No.:G-2640.0001
Specifications
About this item
Customers Who Bought This Also Bought