H-Glu-Asp-OH
Catalog #: G-1910.0250BA
Supplier: Bachem Americas
CAS Number:
Specifications
- CAS No.:3918-84-1
- Size:250 mg
- Storage conditions:–20 ± 5 °C
- MDL No.:MFCD00037308
- Protein/peptide name:H-Glu-Asp-OH
- Molecular weight:262.22
- Sequence:H-Glu-Asp-OH
- Molecular formula:C₉H₁₄N₂O₇
- Cat. no.:G-1910.0250BA
- Supplier No.:G-1910.0250
Specifications
About this item
Customers Who Bought This Also Bought