Cyclo(-Gly-Ser)
Catalog #: G-1725.0001BA
Supplier: Bachem Americas
CAS Number:
Specifications
- CAS No.:52661-98-0
- Size:1 g
- Storage conditions:–20 ± 5 °C
- MDL No.:MFCD00237615
- Protein/peptide name:Cyclo(-Gly-Ser)
- Molecular weight:144.13
- Sequence:Cyclo(-Gly-Ser)
- Molecular formula:C₅H₈N₂O₃
- Cat. no.:G-1725.0001BA
- Supplier No.:G-1725.0001
Specifications
About this item
Also known as dioxopiperazines, piperazine-2,5-diones or DKPs. Diketopiperazines may occur as by-products during peptide synthesis or during the degradation of peptides. These cyclic dipeptides have been detected as taste-modulating compounds in food, they often show biological activity. DKPs are valuable chiral synthons, employed e.g. in Schöllkopf's versatile bislactim ether approach. They also have found use as catalysts for enantioselective synthesis, e.g. in the asymmetric Strecker reaction. See also the TRH metabolite cyclo(-His-Pro), G-1745, and cyclo(-Asp-Phe), G-1695, the major degradation product of aspartame.
Customers Who Bought This Also Bought