H-Asp(Leu-OH)-OH, Bachem
Catalog #: G-1600.0250BA
Supplier: Bachem Americas
CAS Number:
Specifications
- CAS No.:14650-26-1
- Size:250 mg
- Storage conditions:–20 ± 5 °C
- MDL No.:MFCD00057845
- Protein synonyms:H-b-Asp-Leu-OH
- Protein/peptide name:H-Asp(Leu-OH)-OH
- Molecular weight:246.26
- Sequence:H-Asp(Leu-OH)-OH
- Molecular formula:C₁₀H₁₈N₂O₅
- Cat. no.:G-1600.0250BA
- Supplier No.:G-1600.0250
Specifications
About this item
See also G-4750 and β-aspartame (G-3725).
Substrate for E.coli isoaspartyl dipeptidase.
Customers Who Bought This Also Bought