H-Asp(Ala-OH)-OH, Bachem
Catalog #: G-1560.0250BA
Supplier: Bachem Americas
CAS Number:
Specifications
- Size:250 mg
- Storage conditions:–20 ± 5 °C
- MDL No.:MFCD00237832
- Protein synonyms:H-b-Asp-Ala-OH
- Protein/peptide name:H-Asp(Ala-OH)-OH
- Molecular weight:204.18
- Sequence:H-Asp(Ala-OH)-OH
- Molecular formula:C₇H₁₂N₂O₅
- Cat. no.:G-1560.0250BA
- Supplier No.:G-1560.0250
Specifications
About this item
See also G-4750 and β-aspartame (G-3725).
Substrate for bacterial β-aspartylpeptidase.
Customers Who Bought This Also Bought