H-Ala-Asp-OH, Bachem
Catalog #: G-1195.0001BA
Supplier: Bachem Americas
CAS Number:
Specifications
- CAS No.:20727-65-5
- Size:1 g
- Storage conditions:–20 ± 5 °C
- MDL No.:MFCD00038165
- Protein/peptide name:H-Ala-Asp-OH
- Molecular weight:204.18
- Sequence:H-Ala-Asp-OH
- Molecular formula:C₇H₁₂N₂O₅
- Cat. no.:G-1195.0001BA
- Supplier No.:G-1195.0001
Specifications
About this item
Ala-Asp was shown to induce chloride excretion in proximal tubule cells.
Customers Who Bought This Also Bought