Order Entry
ContactUsLinkComponent
CulGenex™ Electrophoresis Buffers, Hardy Diagnostics
CulGenex™ Electrophoresis Buffers, Hardy Diagnostics
Catalog #: CA89405-062
CAS Number:  
TBE Electrophoresis Buffer Powder
CulGenex™ Electrophoresis Buffers, Hardy Diagnostics
Catalog #: CA89405-062
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Packaging:
    Pouch
  • Description:
    TBE (Tris Borate EDTA) Electrophoresis Buffer Powder 10X, 1000 ml pouch
  • Size:
    1000 mL
  • Storage temperature:
    2 to 30 °C
  • Cat. no.:
    CA89405-062
  • Supplier No.:
    C4618

Specifications

About this item

CulGenex™ TAE (Tris Acetate EDTA) Electrophoresis Buffer Powder is recommended for used for agarose gel elctrophoresis in the analyses of nucleic acid products resulting from PCR amplification, purification protocols, or cloning experiments.

CulGenex™ TBE (Tris Borate EDTA) Electrophoresis Buffer Powder is recommended for used for agarose gel elctrophoresis in the analyses of nucleic acid products resulting from PCR amplification, purification protocols, or cloning experiments. CulGenex™ Electrophoresis Buffers are used for agarose gel electrophoresis in the analyses of nucleic acid products resulting from PCR amplification, purification protocols, or cloning experiments.

Customers Who Bought This Also Bought