About this item
SevenExcellence meter has been designed with the objective to optimize the workflow and increase the efficiency of your laboratory work.
- Optimize workflow
- Increase efficiency
- One-click start
- Multiparameter
Just one click is required to start a sample or a series of samples.
USB-stick, LabX direct software data export. TFT display. Interface includes RS232, USB-A, USB-B, LAN, Mini-LTW, Mini-DIN, BNC, Cinch/RCA.
The interface on the instrument allows you to connect peripherals, simultaneously supporting your workflow in the best possible way. SevenExcellence pH/mV meter redefines flexibility and offers various measurement parameters in one meter. It not only measures pH and ORP with classical sensors, but also supports pH measurements with ISFET technology. Starting an analysis, changing the settings and accessing the results is easily achieved thanks to the cleverly-designed touch screen menu.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- CA89261-030
- 75845-216
- CA89261-032
- CA89261-034
- Temperature range:
- –30...130 °C
- –30...130 °C
- –30...130 °C
- –30...130 °C
- Temperature resolution:
- 0.1 °C
- 0.1 °C
- 0.1 °C
- 0.1 °C
- Temperature accuracy:
- ±0.1 °C
- ±0.1 °C
- ±0.1 °C
- ±0.1 °C
- Display:
- TFT
- TFT
- TFT
- TFT
- Data logging capacity:
- 20000 data points, 250 analyses
- 20000 data points, 250 analyses
- 20000 data points, 250 analyses
- 20000 data points, 250 analyses
- Interfaces:
- RS232, USB-A, USB-B, LAN, Mini-LTW, Mini-DIN, BNC, Cinch/RCA
- RS232, USB-A, USB-B, LAN, Mini-LTW, Mini-DIN, BNC, Cinch/RCA
- RS232, USB-A, USB-B, LAN, Mini-LTW, Mini-DIN, BNC, Cinch/RCA
- RS232, USB-A, USB-B, LAN, Mini-LTW, Mini-DIN, BNC, Cinch/RCA
- Parameters:
- pH; ORP
- pH; ORP
- pH; ORP
- pH; ORP