Specifications
- Concentration:0 - 8.0 mg/L Cl₂
- Description:SpecCheck Secondary Gel Standards Set, DPD Chlorine - HR
- Method:DPD
- Instrument:Includes values for all instruments
- Supplier No.:2893300
Specifications
About this item
DPD Chlorine, High Range, 0-6.5 mg/L as CI₂.
-Fast and Convenient
-Stable
- Fast and Convenient
- Stable
DPD Chlorine, High Range, 0-6.5 mg/L as CI₂.
Used to confirm consistent instrument response. Set contains four vials filled with gels that simulate the test color at various concentrations.
*This product has not been evaluated to test for chlorine and chloramines in medical applications in the United States.