Distillation Head, Variable Reflux Ratio, Ace Glass
Catalog #: 89055-692
Supplier: Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
Specifications
- ST Joint No.:24/40, side arm and main joints
- Description:Variable Reflux Ratio Distillation Head
- Hose connection int.Ø:5/16 or 3/8"
- Cat. no.:89055-692
- Supplier No.:6604-10
Specifications
About this item
This variable reflux ratio distillation head is constructed so that satisfactory operation can be maintained under any pressure condition
[ST] 10/30 thermometer joint takes 76 mm immersion thermometer. Fabricated from borosilicate glass.
Customers Who Bought This Also Bought