Order Entry
ContactUsLinkComponent
Stand and Rod Value Pack
Stand and Rod Value Pack
Catalog #: 470137-892
Supplier:  Wards
STAND / ROD 4X6 W/24 ROD WITH 24 ROD
Stand and Rod Value Pack
Catalog #: 470137-892
Supplier:  Wards

Specifications

  • Rod Length:
    508 mm (20")
  • Description:
    Stand and rod value pack
  • Cat. no.:
    470137-892
  • Base Dimensions:
    101×152 mm (4×6")
  • Rod Diameter:
    12 mm (0.47")
  • Supplier No.:
    STDRD

Specifications

About this item

Economically Priced Performer

  • 4×6" base
  • 20" rod

This economically-priced pack is perfect for use in chemistry or physics labs. The base is die-cut, formed steel and has a powder-coated finish.

Customers Who Bought This Also Bought