ContactUsLinkComponent
Air Stirrer, Light Duty, Ace Glass
Air Stirrer, Light Duty, Ace Glass
Catalog #: 89077-660
Supplier:  Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
13365 Air stirrer
Air Stirrer, Light Duty, Ace Glass
Catalog #: 89077-660
Supplier:  Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
Specifications
  • Description:
    Air Stirrer, Light Duty
  • Cat. no.:
    89077-660
  • Supplier No.:
    13365-05
Specifications
About this item
The small, compact, and quiet air motor is ideal for stirring all types of solvents, lacquers, paints, oils, synthetics, and fine and heavy chemicals
The device requires an air supply of only 30 to 100PSI. Variable speed ranges from 200rpm to 10,000rpm simply by turning the air supply line valve. For bath sizes to 20L, this smooth running unit has little or no service cost. Unit adjusts itself to compensate for wear. The shaft and propeller are made of stainless steel to resist most acids and chemicals, and for easier cleaning. The device will start in most stalled positions with low air pressure and cannot burn out from overload.
The Air Motor offers 0–⅓hp at 80lbs. Coupler and stainless steel shaft are 30.5cm (12") overall in length. The propeller is stainless steel, and measures 6.4cm (2½") in diameter. The shaft has a 9.5mm (⅜") O.D. For filter/regulator/lubricator, see 89077-666. Air consumption is 13cfm.
As there are no sparks, the air stirrer virtually eliminates the risk of explosions. It is a complete unit, ready for mounting on a laboratory stand.
Customers Who Bought This Also Bought