Chemical-Duty Dry Diaphragm Vacuum Pumps, High Capacity, Welch-Ilmvac
Supplier: Avantor
Certificates
About this item
Use affordable Welch-Ilmvac dry diaphragm pumps to provide oil-free vacuum for a wide variety of applications
Welch-Ilmvac diaphragm vacuum pumps are used world-wide in industry, laboratory, and education. Economical dry diaphragm pumps are quiet and ecologically superior to oil-lubricated pumps or water aspirator-generated vacuum. For applications requiring rough to medium-high vacuum (1 to 760torr). Use chemical duty pumps with acids, solvent fumes.
Select from a broad range of flow and ultimate vacuum level models to provide reliable, low-maintenance vacuum for routine and specialized needs.
Specifications
- Catalog No:
- 89410-232
- Electrical:
- 115V, 60Hz
- Flow rate:
- 6.1 cfm (173 L/min.)