To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Size:5 mg
- Storage conditions:Freezer
- Protein/peptide name:Z-Asp-Glu-Val-Asp-7-Amido-
- Cat. no.:CAAAJ64569-LB0
- Supplier No.:J64569-LB0
Specifications
About this item
Molecular Formula: C36H38F3N5O14
Formula Weight: 821.71
Storage Temperature: -30°C to -10°C
Physical Form: Powder
Appearance: White to off-white
A fluorogenic substrate for caspase-3
A fluorogenic substrate for caspase-3