Bradykinin (1-3)
Catalog #: CAAAJ61719-LB0
Supplier: Thermo Scientific Chemicals
CAS Number:
Specifications
- Size:5 mg
- Storage conditions:Ambient
- Form:Powder
- Amino acid number:1 to 3
- MDL No.:MFCD00081011
- Protein/peptide name:Bradykinin (1-3)
- Cat. no.:CAAAJ61719-LB0
- Supplier No.:J61719-LB0
Specifications
About this item
Molecular Formula: C16H28N6O4
Formula Weight: 368.40
Physical Form: Powder
Appearance: White
MDL No.: MFCD00081011
Biologically active fragment of bradykinin
Customers Who Bought This Also Bought