Adrenocorticotropic Hormone (1-13)
Supplier: Thermo Scientific Chemicals
Certificates
About this item
CAS No.: 22006-64-0
Molecular Formula: C75H106N20O19S
Formula Weight: 1623.85
Storage Temperature: -30°C to -10°C
Physical Form: Powder
Appearance: White
MDL No.: MFCD00133030
Customers Who Bought This Also Bought