Apamin
Catalog #: CAAAJ60961-EXD
Supplier: Thermo Scientific Chemicals
CAS Number:
Specifications
- CAS No.:24345-16-2
- Size:0.5 mg
- Storage conditions:Freezer
- Form:Powder
- MDL No.:MFCD00167944
- Protein/peptide name:Apamin
- Cat. no.:CAAAJ60961-EXD
- Supplier No.:J60961-EXD
Specifications
About this item
CAS No.: 24345-16-2
Molecular Formula: C79H131N31O24S4
Formula Weight: 2027.37
Storage Temperature: -30°C to -10°C
Physical Form: Powder
Appearance: White to pale brown
Merck Reference: 14,725
MDL No.: MFCD00167944
Neurotoxic peptide isolated from honeybee venom. Blocks calcium-activated potassium channels
Customers Who Bought This Also Bought