To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- CAS No.:57966-42-4
- Size:1mg
- Storage conditions:Freezer
- MDL No.:MFCD00076875
- Protein/peptide name:Proctolin
- Molecular weight:648.762 g/mol
- Molecular formula:C₃₀H₄₈N₈O₈
- Cat. no.:CAAAJ60532-MCR
- Supplier No.:J60532-MCR
Specifications
About this item
An insect neuropeptide.
CAS No.: 57966-42-4 Molecular Formula: C30H48N8O8 Formula Weight: 648.76 Storage Temperature: -30°C to -10°C Physical Form: Powder Appearance: White MDL No.: MFCD00076875
An insect neuropeptide