Clamp Holder, Pilot Plant, Ace Glass
Catalog #: 89075-630
Supplier: Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
Specifications
- Description:Pilot Plant Clamp Holder
- Cat. no.:89075-630
- Supplier No.:11081-21
Specifications
About this item
This clamp is used with versatile jaw-type clamps to secure condensers and other apparatus on pilot plant reactor support frames
The holder clamps to a one-inch bar on the support frame, and the clamp is held in the clamp holder with a thumb screw. Fabricated of aluminum; powder coated black. For clamps, see 89083-252 or 89083-246 (among others).
Customers Who Bought This Also Bought