Clamp, Three-Prong, Vertical, Ace Glass
Catalog #: 89083-210
Supplier: Ace Glass
Small Business Enterprise
Specifications
- Description:Three-Prong Vinylized Jaw Clamp
- Cat. no.:89083-210
Specifications
About this item
This three-prong grip clamp is designed to hold dispensing burets, thermometers, reflux condensers, and other laboratory glassware, in a vertical position
The vinlylized jaws will hold objects up to 57mm in diameter.
Customers Who Bought This Also Bought