Gooseneck Extension Clamps, Ace Glass
Supplier: Ace Glass
Certificates
About this item
This clamping system features a 12" or 16" flex arm
Ideal for working within hoods. The system mounts to any lab frame or support stand with a 19mm or less diameter. An optional base plate or bench clamp provides increased versatility.
Ordering information: Complete units include flex arm, two-prong head, three-prong head, spring head, and lab frame connector. Comes with an extra set of fiberglass prong covers for temperatures above 100°C.
Customers Who Bought This Also Bought