JACKSON SAFETY® Lens Cleaning Towelettes, KIMBERLY-CLARK PROFESSIONAL®
Catalog #: 33300-562
Supplier: Kimberly-Clark
Specifications
- Description:Lens cleaning towelettes
- Cat. no.:33300-562
- Supplier No.:14551
Specifications
About this item
Anti-fog, anti-static formula for cleaning glass, plastic, and polycarbonate lenses, goggles, and visors.
Packaging: 100 individually packaged towelettes per box.
Customers Who Bought This Also Bought