Dura-Clamp® Clamp for Tubing, Ace Glass
Catalog #: 89075-640
Supplier: Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
Specifications
- Description:Tubing Dura-Clamp® Clamp
- Cat. no.:89075-640
- Supplier No.:11146-08
Specifications
About this item
This autoclavable, white plastic unitized construction tubing clamp, can also be used as a flow valve
Instant 15-position control clamp accepts flexible tubing from 3.2 to 11mm O.D. (1/8" to 7/16").
Customers Who Bought This Also Bought