About this item
Kimberly-Clark™ Kimguard™ One-Step™ sterilization wraps are available in six grades, from KC100 to KC600, to meet a full range of applications. The higher the number, the stronger the wrap.
- Fast, easy way to ensure that instrument sterility is uncompromised
- Two layers of wrap, thermally sealed along the sides
- Delivers the protection of double-wrapping in a single step, in half the time it takes to wrap or open packages using sequential double wrapping
- Microbial barrier protection with patented Powerguard™ technology
- Made in USA
- Pre-Vacuum Steam and Ethylene Oxide (EO) compatible
- Flame resistant: Meets NFPA Class 1
This KC400 Kimguard™ One-Step™ wrap is the 6th of 6 grades. Use to package linen towels, packs, gowns, light basin sets, and single instruments for sterilization. Low-lint fabric delivers barrier and strength properties that help prevent contamination after sterilization. Steam and EO compatible. Naturally latex-free. Nonsterile. Multiple sizes.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- CA75819-570
- CA75819-552
- Environmentally Preferable: