About this item
Kimberly-Clark™ Kimguard™ One-Step™ sterilization wraps are available in six grades, from KC100 to KC600, to meet a full range of applications. The higher the number, the stronger the wrap.
- Fast, easy way to ensure that instrument sterility is uncompromised
- Two layers of wrap, thermally sealed along the sides
- Delivers the protection of double-wrapping in a single step, in half the time it takes to wrap or open packages using sequential double wrapping
- Microbial barrier protection with patented Powerguard™ technology
- Made in USA
- Pre-Vacuum Steam and Ethylene Oxide (EO) Compatible
- Flame resistant: Meets NFPA Class 1
This KC500 Kimguard™ One-Step™ wrap is the 5th of 6 grades. Use to package linen towels, packs, gowns, light basin sets, and single instruments for sterilization. Low-lint fabric delivers barrier and strength properties that help prevent contamination after sterilization. Steam and EO compatible. Naturally latex-free. Non sterile. Multiple sizes.