Hydrochloric acid 10% (v/v) (1+9), VWR Chemicals BDH®
Supplier: VWR International
Certificates
About this item
Synonyms: Muriatic acid
Formula:HCl
MW:36.46 g/mol
MDL Number:MFCD00011324
CAS Number:7647-01-0
:Storage Temperature: Ambient
ADR:8,II
:length
:width
:height
:Ø
:thickness
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:M.W. 36.46 g/mol
:36.46
EINECS:231-595-7
UN:1789
:8
:II
:GHS07
Storage Temperature:Ambient
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specification Test Results
- Appearance/color:Clear liquid
- Shelf-life:2 years
- Hydrochloric acid 10% (v/v) (1+9), VWR Chemicals BDH®:
Specification Test Results
Related Information
Certificates
Search by Lot Number20l (5 Gal) Cubitainer
Packaging Information
AT-BDHVBDH7417-1_4_7.pdf spec_BDH7417-1 spec_BDH7417-4 spec_BDH7417-7
Product specification
Avantor Portfolio of Chemicals VWR Top Rated Guide
Product Literature
VWR GHS Compliance Letter (US)
Additional Product Details
Customers Who Bought This Also Bought