Clamps, Stainless Steel, 3-Prong, Ace Glass
Supplier: Ace Glass
Certificates
About this item
This multi-purpose, three-prong clamp is manufactured from electro-polished stainless steel for improved corrosion resistance and autoclave sterilization
Features include dual screw adjustments and vinyl sleeved prongs.
Ordering information: Replacement high temperature fiberglass sleeves and stainless steel clamp holders are available for purchase.
Customers Who Bought This Also Bought