Clamps, Stainless Steel, Ace Glass
Catalog #: 89059-638
Supplier: Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
Specifications
- Description:Stainless Steel Clamps
- Cat. no.:89059-638
- ST Joint Size:For 34/45
- Supplier No.:7600-35
Specifications
About this item
These clamps will securely hold [ST] 10/30 through [ST] 45/50 joints
No screw is needed. Can be used to secure thermometers, vacuum systems, distillation set-ups, chromatographic columns, evaporators, or any glass apparatus that has to be secured with springs or clamps. These clamps are made to last, of corrosion-free, stainless steel spring wire.
Customers Who Bought This Also Bought