To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- CAS No.:189950-66-1
- Size:25 mg
- Storage conditions:Freezer
- MDL No.:MFCD00792707
- Protein/peptide name:N-Acetyl-Asp-Glu-Val-Asp p-nitroanilide
- Molecular weight:638.58 g/mol
- Molecular formula:C₂₆H₃₄N₆O₁₃
- Cat. no.:CAAAJ65878-MB
- Supplier No.:J65878-MB
Specifications
About this item
A colorimetric substrate for caspase-3.
CAS No.: 189950-66-1 Molecular Formula: C26H34N6O13 Formula Weight: 638.58 Storage Temperature: -30°C to -10°C Physical Form: Powder Appearance: White MDL No.: MFCD00792707
Applications: A colorimetric substrate for caspase-3