Factor XA protease digestion buffer, Thermo Scientific
Catalog #: CAAAJ63878-AK
Supplier: Thermo Scientific Chemicals
CAS Number:
Specifications
- Size:250 mL
- Cat. no.:CAAAJ63878-AK
- Supplier No.:J63878-AK
Specifications
About this item
Liquid
Physical Form: Liquid
Applications: For His-tag protein purification from mammalian cells
Customers Who Bought This Also Bought