Tris(hydroxymethyl)aminomethane Hydrochloride, 1M stock soln, pH 8.0, Thermo Scientific
Catalog #: CAAAJ63636-K2
Supplier: Thermo Scientific Chemicals
CAS Number:
Specifications
- Size:1 L
- MDL No.:MFCD00012590
- Cat. no.:CAAAJ63636-K2
- Supplier No.:J63636-K2
Specifications
About this item
Liquid
CAS No.: 1185-53-1
Physical Form: Liquid
Appearance: Colorless
MDL No.: MFCD00012590
Customers Who Bought This Also Bought