Phosphate-Citrate Buffer (5X), Thermo Scientific
Catalog #: CAAAJ62982-AE
Supplier: Thermo Scientific Chemicals
CAS Number:
Specifications
- pH:2.5
- Description:Phosphate-citrate buffer (5X)
- Size:100 mL
- MDL No.:MFCD00131859
- Cat. no.:CAAAJ62982-AE
- Supplier No.:J62982-AE
Specifications
About this item
Phosphate-Citrate Buffer (5X) is used as a substrate buffer in immunoassay procedures. It is used in SDS PAGE electrophoresis.
Contains 1M sodium phosphate and 0.5M sodium citrate.
Customers Who Bought This Also Bought