Tris(hydroxymethyl)aminomethane hydrochloride, 1M soln., pH 7,4, RNase free, Thermo Scientific
Catalog #: CAAAJ62848-AK
Supplier: Thermo Scientific Chemicals
CAS Number:
Specifications
- Size:250 mL
- MDL No.:MFCD00012590
- Cat. no.:CAAAJ62848-AK
- Supplier No.:J62848-AK
Specifications
About this item
Liquid
CAS No.: 1185-53-1
Physical Form: Liquid
Appearance: Colorless
MDL No.: MFCD00012590
Customers Who Bought This Also Bought