bisBenzimide H-33342 trihydrochloride trihydrate (Hoechst 33342) 98%
Catalog #: CAAAJ62134-MC
Supplier: Thermo Scientific Chemicals
CAS Number: 23491-52-3
Specifications
- Size:100 mg
- Cat. no.:CAAAJ62134-MC
- Supplier No.:J62134-MC
Specifications
About this item
Synonyms: Hoechst 33342,2′-(4-Ethoxyphenyl)-5-(4-methyl-1-piperazinyl)-2,5′-bi-1H-benzimidazole trihydrochloride,HOE 33342
:length
:width
:height
:Ø
:thickness
:98%
:97,5
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:M.W. 615.99 g/mol (561.94 anhy)
Formula:C₂₇H₂₈N₆O·3HCl·3H₂O
MW:615.99 g/mol (561.94 anhy)
Storage Temperature:Refrigerator
MDL Number:MFCD00012679
CAS Number:23491-52-3
EINECS:245-691-1
Customers Who Bought This Also Bought