Laemmli SDS sample buffer, reducing (6X), Thermo Scientific
Supplier: Thermo Scientific Chemicals
Certificates
About this item
Liquid
Physical Form: Liquid
Appearance: Blue
Applications: For protein sample preparation to be used in the Laemmli SDS-PAGE system
Related Information
Customers Who Bought This Also Bought