4-Aminobutyric acid 99+%
Catalog #: CAAAJ61307-09
Supplier: Thermo Scientific Chemicals
CAS Number: 56-12-2
Specifications
- Size:10 g
- Cat. no.:CAAAJ61307-09
- Supplier No.:J61307-09
Specifications
About this item
Synonyms: GABA
:4-Aminobutyric acid
:Storage Temperature: Ambient
Boiling Pt:248 °C (1013 hPa)
Melting Pt:∼ 195 °C (dec.)
Density:1.21 g/cm³ (20 °C)
:length
:width
:height
:Ø
:thickness
:99+%
:99,0
:Boiling Pt:
:Boiling Pt: 248 °C (1013 hPa)
:Melting Pt:
:Melting Pt: ∼ 195 °C (dec.)
:Flash Pt:
:Flash Pt:
:M.W. 103.12 g/mol
Merck Index:13,00429
Formula:C₄H₉NO₂
MW:103.12 g/mol
Storage Temperature:Ambient
EINECS:200-258-6
:∼ 195 °C
:248 °C (1013 hPa)
:14,430
MDL Number:MFCD00008226
CAS Number:56-12-2
Customers Who Bought This Also Bought